Få mere at vide
Tocris Bioscience™ IRAK1/4 Inhibitor I
IRAK4 and IRAK1 inhibitor
Brand:  Tocris Bioscience™ 5665/50
Encompass
Produktkode 15767026
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
IRAK1/4 Inhibitor I is an IRAK4 and IRAK1 inhibitor (IC50 values are 0.2 and 0.3 μM, respectively).
Tekniske data
Tekniske data
N-[1-[2-(4-Morpholinyl)ethyl]-1H-benzimidazol-2-yl]-3-nitrobenzamide
509093-47-4