CAS RN 508169-18-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
7-Chloromethyloxy-carbonyloxy-6-(5-chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-Chloromethyloxy-carbonyloxy-6-(5-chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-Chloromethyloxy-carbonyloxy-6-(5-chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.