CAS RN 5063-65-0
IUPAC Navn:
2-pentyloxirane
Synonymer:
2-pentyloxirane
Molekylvægt (g/mol):
114.19
Molekylformel:
C7H14O
InChi Key:
NMOFYYYCFRVWBK-UHFFFAOYNA-N
SMILES:
CCCCCC1CO1
1
–
3
af
3
Resultater
1
1,2-Epoxyheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2-Epoxyheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2-Epoxyheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.