CAS RN 50508-73-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
[2-[[3-(2-Chlorobenzoyl)-5-ethyl-2-thienyl]amino]-2-oxoethyl]carbamic Acid Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
[2-[[3-(2-Chlorobenzoyl)-5-ethyl-2-thienyl]amino]-2-oxoethyl]carbamic Acid Benzyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.