CAS RN 504-30-3
IUPAC Navn:
2,3-dihydropyridazin-3-one
Synonymer:
2H-pyridazin-3-one
Molekylvægt (g/mol):
96.09
Molekylformel:
C4H4N2O
InChi Key:
AAILEWXSEQLMNI-UHFFFAOYSA-N
SMILES:
O=C1NN=CC=C1
1
–
3
af
3
Resultater
1
Pyridazin-3(2H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pyridazin-3(2H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pyridazin-3(2H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.