CAS RN 502487-67-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SQ109, MedChemExpress
MedChemExpress SQ109 is a potent inhibitor of the trypomastigote form of the parasite, with IC50 for cell killing of 50±8 nM. SQ109, targets MmpL3, is an antitubercular agent.