Få mere at vide
GANT 61, MedChemExpress
Artikelnummer. 16495898
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16495898 10 mg -
16482047 50 mg -
16478537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16495898 Leverandør Medchem Express Leverandørnr. HY13901
MedChemExpress GANT 61 is an inhibitor of Gli1 and Gli2 targeting the Hedgehog/GLI pathway.
GANT 61 is an inhibitor of Gli1 and Gli2 targeting the Hedgehog/GLI pathway.
Tekniske data
Kemisk navn eller materiale GANT 61
Fysisk form Solid
CAS 500579-04-4
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C27H35N5
Til brug med (applikation) Cancer-programmed cell death
Synonym NSC 136476
Opløselighedsinformation Ethanol : 66.67 mg/mL (155.19 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (58.19 mM; Need ultrasonic)
SMIL CN(C)C1=CC=CC=C1CN2C(C3=CC=NC=C3)N(CC4=CC=CC=C4N(C)C)CCC2
Molekylvægt (g/mol) 429.6
Formel vægt 429.6
Procent renhed 98.0%