CAS RN 4986-89-4
IUPAC Navn:
3-(prop-2-enoyloxy)-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate
Synonymer:
pentaerythritol tetraacrylate
Molekylvægt (g/mol):
352.34
Molekylformel:
C17H20O8
InChi Key:
KNSXNCFKSZZHEA-UHFFFAOYSA-N
SMILES:
C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
1
–
3
af
3
Resultater
1
Pentaerythritol Tetraacrylate (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pentaerythritol Tetraacrylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pentaerythritol Tetraacrylate (~80%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.