CAS RN 49705-98-8
IUPAC Navn:
(1R,2S)-1-benzyl-1-carbamoyl-2-hydroxypropyl carbamate
Synonymer:
(1R,2S)-1-benzyl-1-carbamoyl-2-hydroxypropyl carbamate
Molekylvægt (g/mol):
252.27
Molekylformel:
C12H16N2O4
InChi Key:
XYJIXTMTYOGXGG-QPUJVOFHSA-N
SMILES:
C[C@H](O)[C@@](CC1=CC=CC=C1)(OC(N)=O)C(N)=O
1
–
3
af
3
Resultater
1
N-Benzyloxycarbonyl L-Threonine Amide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Benzyloxycarbonyl L-Threonine Amide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Benzyloxycarbonyl L-Threonine Amide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.