Få mere at vide
N-Benzyloxycarbonyl L-Threonine Amide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Benzyloxycarbonyl Threonine Amide |
| CAS | 49705-98-8 |
| Molekylær formel | C12 H16 N2 O4 |
| Mængde | 1 g |
| Synonym | Carbamic acid, N-[(1S,2R)-1-(aminocarbonyl)-2-hydroxypropyl]-, phenylmethyl ester, Carbamic acid, [(1S,2R)-1-(aminocarbonyl)-2-hydroxypropyl]-, phenylmethyl ester (9CI), Carbamic acid, [1-(aminocarbonyl)-2-hydroxypropyl]-, phenylmethyl ester, [R-(R*,S*)]-, N-(Benzyloxycarbonyl)-L-threonine amide, N-Benzyloxycarbonylthreonine amide |
| InChI formel | InChI=1S/C12H16N2O4/c1-8(15)10(11(13)16)14-12(17)18-7-9-5-3-2-4-6-9/h2-6,8,10,15H,7H2,1H3,(H2,13,16)(H,14,17)/t8-,10+/m1/s1 |
| SMIL | C[C@@H](O)[C@H](NC(=O)OCc1ccccc1)C(=O)N |
| IUPAC navn | benzyl N-[(2S,3R)-1-amino-3-hydroxy-1-oxobutan-2-yl]carbamate |
| Molekylvægt (g/mol) | 252.27 |
| Formel vægt | 252.111 |