CAS RN 4955-90-2
IUPAC Navn:
sodium 2,5-dihydroxybenzoate
Synonymer:
sodium gentisate 龙胆酸钠
Molekylvægt (g/mol):
176.1
Molekylformel:
C7H5NaO4
InChi Key:
MOIJZWWOFOQFMH-UHFFFAOYSA-M
SMILES:
[Na+].OC1=CC=C(O)C(=C1)C([O-])=O
1
–
2
af
2
Resultater
1
Gentisic Acid Sodium Hydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Gentisic Acid Sodium Hydrate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.