CAS RN 49548-40-5
IUPAC Navn:
benzyl N-(carbamothioylmethyl)carbamate
Synonymer:
benzyl N-(carbamothioylmethyl)carbamate
Molekylvægt (g/mol):
224.28
Molekylformel:
C10H12N2O2S
InChi Key:
NWHXDCPLGZXNOR-UHFFFAOYSA-N
SMILES:
NC(=S)CNC(=O)OCC1=CC=CC=C1
1
–
3
af
3
Resultater
1
N-Benzyloxycarbonylglycine Thioamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Benzyloxycarbonylglycine Thioamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Benzyloxycarbonylglycine Thioamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.