CAS RN 4928-87-4
IUPAC Navn:
1H-1,2,4-triazole-5-carboxylic acid
Synonymer:
2H-1,2,4-triazole-3-carboxylic acid
Molekylvægt (g/mol):
113.08
Molekylformel:
C3H3N3O2
InChi Key:
LJVQHXICFCZRJN-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NC=NN1
1
–
1
af
1
Resultater
1
1H-1,2,4-Triazole-3-carboxylic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.