CAS RN 486460-00-8
IUPAC Navn:
3-{[(tert-butoxy)carbonyl]amino}-4-(2,4,5-trifluorophenyl)butanoic acid
Synonymer:
3-[(tert-butoxycarbonyl)amino]-4-(2,4,5-trifluorophenyl)butanoic acid
Molekylvægt (g/mol):
333.31
Molekylformel:
C15H18F3NO4
InChi Key:
TUAXCHGULMWHIO-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CC(O)=O)CC1=CC(F)=C(F)C=C1F
1
–
4
af
4
Resultater
1
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3R)-3-(tert-Butoxycarbonylamino)-4-(2,4,5-trifluorophenyl)butanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
(R)-3-((tert-Butoxycarbonyl)amino)-4-(2,4,5-trifluorophenyl)butanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.