CAS RN 479-41-4
IUPAC Navn:
(E)-1',2'-dihydro-1H,3H-[2,3'-biindolylidene]-2',3-dione
Synonymer:
indirubin
Molekylvægt (g/mol):
262.27
Molekylformel:
C16H10N2O2
InChi Key:
CRDNMYFJWFXOCH-BUHFOSPRSA-N
SMILES:
O=C1NC2=CC=CC=C2\C1=C1/NC2=CC=CC=C2C1=O
1
–
4
af
4
Resultater
1
Indigopurpurin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Indigopurpurin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Indigopurpurin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Indirubin, MedChemExpress
MedChemExpress Indirubin (Couroupitine B) is a bis-indole alkaloid and has emarkable anticancer activity against chronic myelocytic leukemia.