CAS RN 4737-50-2
IUPAC Navn:
pentylboronic acid
Synonymer:
pentylboronic acid
Molekylvægt (g/mol):
115.97
Molekylformel:
C5H13BO2
InChi Key:
ABWPXVJNCQKYDR-UHFFFAOYSA-N
SMILES:
CCCCCB(O)O
1
–
3
af
3
Resultater
1
Pentylboronic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Pentylboronic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Pentylboronic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.