CAS RN 467458-02-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Idalopirdine Hydrochloride, MedChemExpress
MedChemExpress Idalopirdine Hydrochloride (Lu AE58054 Hydrochloride) is a potent and selective 5-HT6 receptor antagonist with a Ki of 0.83 nM.