CAS RN 455264-30-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Zaurategrast ethyl ester, MedChemExpress
MedChemExpress Zaurategrast ethyl ester (CDP323), the ethyl ester prodrug of CT7758, is a α4β1/α4β7 integrin antagonist used for the treatment of inflammatory and autoimmune disorders.