CAS RN 4549-31-9
IUPAC Navn:
1,7-dibromoheptane
Synonymer:
heptane, 1,7-dibromo-
Molekylvægt (g/mol):
258
Molekylformel:
C7H14Br2
InChi Key:
LVWSZGCVEZRFBT-UHFFFAOYSA-N
SMILES:
BrCCCCCCCBr
1
–
3
af
3
Resultater
1
1,7-Dibromoheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,7-Dibromoheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,7-Dibromoheptane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.