CAS RN 448211-43-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4,4,5,5-Tetramethyl-2-(2-methyl-1-cyclohexen-1-yl)-1,3,2-dioxaborolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,4,5,5-Tetramethyl-2-(2-methyl-1-cyclohexen-1-yl)-1,3,2-dioxaborolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,4,5,5-Tetramethyl-2-(2-methyl-1-cyclohexen-1-yl)-1,3,2-dioxaborolane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.