CAS RN 4461-39-6
IUPAC Navn:
(3-azaniumylpropyl)(2-hydroxyethyl)azanium
Synonymer:
(3-ammoniopropyl)(2-hydroxyethyl)azanium
Molekylvægt (g/mol):
120.2
Molekylformel:
C5H16N2O
InChi Key:
GHKSKVKCKMGRDU-UHFFFAOYSA-P
SMILES:
[NH3+]CCC[NH2+]CCO
1
–
3
af
3
Resultater
1
2-(3-Aminopropylamino)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(3-Aminopropylamino)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(3-Aminopropylamino)ethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.