CAS RN 445-28-3
IUPAC Navn:
2-fluorobenzamide
Synonymer:
2-fluorobenzamide
Molekylvægt (g/mol):
139.13
Molekylformel:
C7H6FNO
InChi Key:
KGGHWIKBOIQEAJ-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC=CC=C1F
1
–
3
af
3
Resultater
1
2-Fluorobenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Fluorobenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Fluorobenzamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.