CAS RN 442851-50-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
3-Dehydroxy 2,3-Dehydro Atorvastatin Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-[2-[(2S)-3,6-Dihydro-6-oxo-2H-pyran-2-yl]ethyl]-5-(4-fluorophenyl)-2-(1-methylethyl)-N,4-diphenyl-1H-pyrrole-3-carboxamide (Atorvastatin eliminated Lactone), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
3-Dehydroxy 2,3-Dehydro Atorvastatin Lactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.