CAS RN 442666-98-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Anguizole, MedChemExpress
MedChemExpress Anguizole is a small molecule inhibitor of HCV replication and alters NS4B(acute)s subcellular distribution.