CAS RN 4424-20-8
IUPAC Navn:
2,3,4,5-tetrahydro-1H-3-benzazepine
Synonymer:
2,3,4,5-tetrahydro-1H-3-benzazepine
Molekylvægt (g/mol):
147.22
Molekylformel:
C10H13N
InChi Key:
MWVMYAWMFTVYED-UHFFFAOYSA-N
SMILES:
C1CC2=CC=CC=C2CCN1
1
–
1
af
1
Resultater
1
5-Azabenzocycloheptene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.