Få mere at vide
5-Azabenzocycloheptene, TRC
Artikelnummer. 30347077
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30347077 500 mg
30346315 1 g
30338069 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30347077 Leverandør Toronto Research Chemicals Leverandørnr. TRCA795190500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Azabenzocycloheptene
CAS 4424-20-8
Molekylær formel C10H13N
Mængde 500 mg
Synonym 1,2,4,5-Tetrahydro-3H-3-benzazepine, 2,3,4,5-Tetrahydro-1H-3-benzazepine, 2,3,4,5-Tetrahydro-1H-benzo[d]azepine
InChI formel InChI=1S/C10H13N/c1-2-4-10-6-8-11-7-5-9(10)3-1/h1-4,11H,5-8H2
SMIL C1Cc2ccccc2CCN1
IUPAC navn 2,3,4,5-tetrahydro-1H-3-benzazepine
Molekylvægt (g/mol) 147.22
Formel vægt 147.1048