CAS RN 432-60-0
IUPAC Navn:
(1R,3aS,3bR,9aR,9bS,11aS)-11a-methyl-1-(prop-2-en-1-yl)-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonymer:
allylestrenol allylestrenol (substance) gestanol gestanon gestanin allilestrenolo gestanyn organon brand of allylestrenol allyloestrenol - chemical alilestrenol
Molekylvægt (g/mol):
300.49
Molekylformel:
C21H32O
InChi Key:
ATXHVCQZZJYMCF-XUDSTZEESA-N
SMILES:
C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)CC=C
1
–
2
af
2
Resultater
1
Allylestrenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Allylestrenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.