CAS RN 42588-37-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Canada Pesticide Mixture 6 ver. 2 500-2000 μg/mL in Toluene:Isooctane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Enstar, TRC
CAS: 42588-37-4 Molekylær formel: C18 H28 O2 Molekylvægt (g/mol): 276.41 Synonym: 2,4-Dodecadienoic acid, 3,7,11-trimethyl-, 2-propynyl ester, (2E,4E)- (9CI),2,4-Dodecadienoic acid, 3,7,11-trimethyl-, 2-propynyl ester, (E,E)-,ENT 70531,Enstar,Kinoprene,XR 777,ZR 77 IUPAC navn: prop-2-ynyl (2E,4E)-3,7,11-trimethyldodeca-2,4-dienoate SMIL: CC(C)CCCC(C)C\C=C\C(=C\C(=O)OCC#C)\C