CAS RN 415724-02-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
1,3-Bis(2,5-dimethylphenoxy)propane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,3-Bis(2,5-dimethylphenoxy)propane, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
1,3-Bis(2,5-dimethylphenoxy)propane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
1,3-Bis(2,5-dimethylphenoxy)propane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.