Få mere at vide
1,3-Bis(2,5-dimethylphenoxy)propane, Mikromol
Artikelnummer. 30458500
Skift visning
Mængde:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458500 4 x 25 mg
30051828 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30458500 Leverandør Mikromol Leverandørnr. MM0641.0200254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,3-Bis(2,5-dimethylphenoxy)propane, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Bis(2,5-dimethylphenoxy)propane
CAS 415724-02-6
Analyt- eller komponentnavne 1,3-Bis(2,5-dimethylphenoxy)propane
Forsendelsestilstand Room Temperature
Molekylær formel C19 H24 O2
Certificeringer/compliance ISO 9001
Synonym Gemfibrozil Imp. H (EP), Benzene, 1,1'-[1,3-propanediylbis(oxy)]bis[2,5-dimethyl- (9CI), 1,3-Bis(2,5-dimethylphenoxy)propane, Gemfibrozil Impurity H
InChI formel InChI=1S/C19H24O2/c1-14-6-8-16(3)18(12-14)20-10-5-11-21-19-13-15(2)7-9-17(19)4/h6-9,12-13H,5,10-11H2,1-4H3
Løsningstype Neat
SMIL Cc1ccc(C)c(OCCCOc2cc(C)ccc2C)c1
IUPAC navn 2-[3-(2,5-dimethylphenoxy)propoxy]-1,4-dimethylbenzene
Molekylvægt (g/mol) 284.39
Mængde 4 x 25 mg
Formel vægt 284.1776 g/mol