CAS RN 41489-89-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Salbutamon Hydrochloride, TRC
CAS: 41489-89-8 Molekylær formel: C13 H19 N O3 . Cl H Molekylvægt (g/mol): 273.76 Synonym: 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride,Salbutamol Sulphate Imp. J (EP) as Hydrochloride,Salbutamone Hydrochloride IUPAC navn: 2-(tert-butylamino)-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone;hydrochloride SMIL: Cl.CC(C)(C)NCC(=O)c1ccc(O)c(CO)c1
2
2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride (Salbutamone Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.