Få mere at vide
2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride (Salbutamone Hydrochloride), Mikromol™
Beskrivelse
2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride (Salbutamone Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride (Salbutamone Hydrochloride) |
| CAS | 41489-89-8 |
| Analyt- eller komponentnavne | 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride (Salbutamone Hydrochloride) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C13 H19 N O3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-[(1,1-Dimethylethyl)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride, Salbutamol Sulphate Imp. J (EP) as Hydrochloride, Salbutamone Hydrochloride, AH 3906A, Ph Eur Salbutamol Impurity J, Salbutamol ketone, Albuterol Related Compound B as Hydrocloride as Hydrochloride, Salbutamol Impurity J as Hydrochloride, Salbutamol Sulfate Impurity J as Hydrochloride, Salbutamol Impurity J as Hydrochloride |
| InChI formel | InChI=1S/C13H19NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,14-16H,7-8H2,1-3H3;1H |
| SMIL | Cl.CC(C)(C)NCC(=O)c1ccc(O)c(CO)c1 |
| IUPAC navn | 2-(tert-butylamino)-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone;hydrochloride |
| Vis mere |