CAS RN 413611-93-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
10074-G5, MedChemExpress
MedChemExpress 10074-G5 is an inhibitor of c-Myc-Max dimerization with an IC50 of 146 μM.