CAS RN 4128-17-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
(2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-yl Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-yl Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-yl Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.