CAS RN 4037-01-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human is a melanocortin 4 (MC4R) receptor agonist.
2
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human is a melanocortin 4 (MC4R) receptor agonist.
3
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human
MedChemExpress Adrenocorticotropic Hormone (ACTH) (4-10), human is a melanocortin 4 (MC4R) receptor agonist.