CAS RN 40013-87-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
24, 25-Dihydroxy VD3, MedChemExpress
MedChemExpress 24, 25-Dihydroxy VD3 is a compound closely related to 1,25-dihydroxyvitamin D3 and is the active form of vitamin D3.