CAS RN 3952-98-5
IUPAC Navn:
potassium (E)-(1-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]sulfanyl}but-3-en-1-ylidene)amino sulfate
Synonymer:
sinigrin
Molekylvægt (g/mol):
397.45
Molekylformel:
C10H16KNO9S2
InChi Key:
QKFAFSGJTMHRRY-OCFLFPRFSA-M
SMILES:
[K+].OC[C@H]1O[C@@H](S\C(CC=C)=N\OS([O-])(=O)=O)[C@H](O)[C@@H](O)[C@@H]1O
1
–
3
af
3
Resultater
1
Sinigrine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Sinigrine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Sinigrine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.