CAS RN 393105-53-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
5
af
5
Resultater
1
Tiplaxtinin, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tocris Bioscience™ Tiplaxtinin
Plasminogen activator inhibitor-1 (PAI-1) inhibitor
3
Tiplaxtinin, MedChemExpress
MedChemExpress Tiplaxtinin is a selective and orally efficacious inhibitor of plasminogen activator inhibitor-1 (PAI-1) with IC50 of 2.7 μM.
4
5