Få mere at vide
Tocris Bioscience™ Tiplaxtinin
Plasminogen activator inhibitor-1 (PAI-1) inhibitor
Brand: Tocris Bioscience™ 5565/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Tiplaxtinin is a plasminogen activator inhibitor-1 (PAI-1) inhibitor (IC50 = 2.7 μM). Induces vascular smooth muscle cell apoptosis in vitro and reduces carotid artery neointimal formation in a rat vascular injury model. Promotes thrombolysis in rat and dog models of acute arterial thrombosis. Orally available.Tekniske data
| α-Oxo-1-(phenylmethyl)-5-[4-(trifluoromethoxy)phenyl]-1H-indole-3-acetic acid | |
| 393105-53-8 | |
| PAI 039, tiplasinin | |
| 50 mg |