CAS RN 38850-51-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N,N,N-Trimethyl-3-[[(tridecafluorohexyl)sulfonyl]amino]-1-propanaminium, Inner Salt(>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N,N-Trimethyl-3-[[(tridecafluorohexyl)sulfonyl]amino]-1-propanaminium, Inner Salt(>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N,N,N-Trimethyl-3-[[(tridecafluorohexyl)sulfonyl]amino]-1-propanaminium, Inner Salt(>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.