CAS RN 38579-27-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
MedChemExpress H-Lys-Trp-Lys-OH
MedChemExpress H-Lys-Trp-Lys-OH is a small molecule peptide which displays antibacterial and antiviral activities extracted from patent CN 104072579 A, Compound AMP12.
2
MedChemExpress H-Lys-Trp-Lys-OH
MedChemExpress H-Lys-Trp-Lys-OH is a small molecule peptide which displays antibacterial and antiviral activities extracted from patent CN 104072579 A, Compound AMP12.
3
MedChemExpress H-Lys-Trp-Lys-OH
MedChemExpress H-Lys-Trp-Lys-OH is a small molecule peptide which displays antibacterial and antiviral activities extracted from patent CN 104072579 A, Compound AMP12.