CAS RN 38385-95-4
IUPAC Navn:
2-(piperidin-4-yl)-1H-1,3-benzodiazole
Synonymer:
2-(piperidin-4-yl)-1H-1,3-benzodiazole
Molekylvægt (g/mol):
201.27
Molekylformel:
C12H15N3
InChi Key:
HBOGHPAOOWUTLB-UHFFFAOYSA-N
SMILES:
C1CC(CCN1)C1=NC2=CC=CC=C2N1
1
–
3
af
3
Resultater
1
2-Piperidin-4-yl-1H-benzimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Piperidin-4-yl-1H-benzimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Piperidin-4-yl-1H-benzimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.