CAS RN 36357-38-7
IUPAC Navn:
1-(6-methylpyridin-3-yl)ethan-1-one
Synonymer:
1-(6-methylpyridin-3-yl)ethanone
Molekylvægt (g/mol):
135.17
Molekylformel:
C8H9NO
InChi Key:
PVRYOKQFLBSILA-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(C)N=C1
1
–
1
af
1
Resultater
1
3-Acetyl-6-methylpyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.