CAS RN 36304-84-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Dimemorfan phosphate, MedChemExpress
MedChemExpress Dimemorfan phosphate is a sigma 1 receptor agonist, used as a potent antitussive.