CAS RN 3601-66-9
IUPAC Navn:
2-{[(tert-butoxy)carbonyl]amino}-2-phenylacetic acid
Synonymer:
[(tert-butoxycarbonyl)amino](phenyl)acetic acid
Molekylvægt (g/mol):
251.28
Molekylformel:
C13H17NO4
InChi Key:
HOBFSNNENNQQIU-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
N-Boc-DL-phenylglycin, 98 %, Thermo Scientific Chemicals
CAS: 3601-66-9 Molekylær formel: C13H17NO4 Molekylvægt (g/mol): 251.282 MDL nummer: MFCD00971063 InChI nøgle: HOBFSNNENNQQIU-UHFFFAOYSA-N Synonym: boc-dl-phg-oh,tert-butoxycarbonylamino-phenyl-acetic acid,2-tert-butoxycarbonyl amino-2-phenylacetic acid,tert-butoxycarbonyl amino phenyl acetic acid,n-boc-dl-phenylglycine,boc-phenylglycine,2-tert-butoxy carbonyl amino-2-phenylacetic acid,rarechem ak ml 0503,2-tert-butoxycarbonylamino-2-phenylacetic acid,boc-d-alpha-phenylglycine PubChem CID: 571805 IUPAC navn: 2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenyleddikesyre SMIL: CC(C)(C)OC(=O)NC(C1=CC=CC=C1)C(=O)O