CAS RN 35900-26-6
IUPAC Navn:
4-methylnonan-5-one
Synonymer:
4-methylnonan-5-one
Molekylvægt (g/mol):
156.27
Molekylformel:
C10H20O
InChi Key:
CGHJMKONNFWXHO-UHFFFAOYNA-N
SMILES:
CCCCC(=O)C(C)CCC
1
–
1
af
1
Resultater
1
4-Methyl-5-nonanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.