CAS RN 35858-81-2
IUPAC Navn:
benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Synonymer:
benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Molekylvægt (g/mol):
329.8
Molekylformel:
C18H18ClN2O2
InChi Key:
BYXCCQMIJIPTSL-UHFFFAOYNA-N
SMILES:
[Cl].NC(CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1
1
–
2
af
2
Resultater
1
L-Tryptophan Benzyl Ester Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
L-Tryptophan benzyl ester hydrochloride, 98%, Thermo Scientific™
CAS: 35858-81-2 Molekylær formel: C18H18ClN2O2 Molekylvægt (g/mol): 329.80 MDL nummer: MFCD00058122 InChI nøgle: BYXCCQMIJIPTSL-UHFFFAOYNA-N Synonym: h-trp-obzl.hcl,l-tryptophan benzyl ester hydrochloride,benzyl l-tryptophanate hydrochloride,h-trp-obzl hcl,h-trp-obzl hydrochloride,s-benzyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride,h-trp-obzlhcl,benzyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride,benzyl l-tryptophanate monohydrochloride,c18h18n2o2.hcl PubChem CID: 45073232 SMIL: [Cl].NC(CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1