CAS RN 35845-62-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Hydrocinnamic-d5 Acid (phenyl-d5), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Hydrocinnamic-d5 Acid (phenyl-d5), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
Hydrocinnamic-d5 Acid (phenyl-d5), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.