CAS RN 35256-85-0
IUPAC Navn:
N-benzyl-2,2-dimethyl-N-(propan-2-yl)propanamide
Synonymer:
tebutam
Molekylvægt (g/mol):
233.36
Molekylformel:
C15H23NO
InChi Key:
RJKCKKDSSSRYCB-UHFFFAOYSA-N
SMILES:
CC(C)N(CC1=CC=CC=C1)C(=O)C(C)(C)C
1
–
3
af
3
Resultater
1
Tebutam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tebutam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tebutam, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.