CAS RN 34896-80-5
IUPAC Navn:
1-bromo-3-methanesulfonylbenzene
Synonymer:
1-bromo-3-methanesulfonylbenzene
Molekylvægt (g/mol):
235.1
Molekylformel:
C7H7BrO2S
InChi Key:
WBOMXUMQOVQNKT-UHFFFAOYSA-N
SMILES:
CS(=O)(=O)C1=CC=CC(Br)=C1
1
–
3
af
3
Resultater
1
3-Bromophenylmethyl Sulfone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Bromophenylmethyl Sulfone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Bromophenylmethyl Sulfone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.